Anti-Connexin 40/GJA5 Rabbit Polyclonal Antibody
BSBTPA1368
- Antibody type:Primary
- Antigen name:Connexin 40
- Antigen symbol:Cx40
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:C-Terminal
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:GJA5
- Amino acid number:341 - 358
- Sequence:KRRLSKASSKARSDDLSV
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human Connexin 40/GJA5 (341-358aa KRRLSKASSKARSDDLSV), identical to the related mouse and rat sequences.
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Rabbit IgG polyclonal antibody for Gap junction alpha-5 protein(GJA5) detection. Tested with WB, IHC-P in Human; Mouse; Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: Connexin 40/GJA5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat