Anti-TNF beta Rabbit Polyclonal Antibody
Catalog # BSBTPA1361
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Tumer Necrosis Factor Beta
- Antigen symbol:TNFB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:TNF b|TNF-b|TNFbeta|tumor necrosis factor|Lymphotoxin-alpha|TNF superfamily member 2|tnf|tumor necorsis factor beta
- Amino acid number:73 - 88
- Sequence:KQNSLLWRANTDRAFL
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence in the middle region of human TNF beta (73-88aa KQNSLLWRANTDRAFL), different from the related mouse sequence by one amino acid.
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for Lymphotoxin-alpha(LTA) detection. Tested with WB in Human.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: TNF beta
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human