Anti-uPA Receptor Rabbit Polyclonal Antibody
BSBTPA1344
- Antibody type:Primary
- Antigen name:Urokinase plasminogen activator receptor
- Antigen symbol:UPAR
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:C-Terminal
- Cross adsorption:No
- Form:Lyophilised
- Antigen synonyms:PLAUR|URKR|uPAR|MO3|Monocyte activation antigen Mo3|Urokinase plasminogen activator surface receptor|CD87|UPAR|U-PAR|CD_antigen|Urokinase plasminogen activator receptor
- Amino acid number:293 - 304
- Sequence:CNHPDLDVQYRS
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human uPA Receptor (293-304aa CNHPDLDVQYRS)
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Rabbit IgG polyclonal antibody for Urokinase plasminogen activator surface receptor (PLAUR) detection. Tested with WB, IHC-P in Human.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: uPA Receptor
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human