Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-S100 beta Rabbit Polyclonal Antibody
Anti-S100 beta Rabbit Polyclonal Antibody
Catalog # BSBTPA1303
Supplier:  Boster Bio
undefined
Anti-S100 beta Rabbit Polyclonal Antibody
Catalog # BSBTPA1303
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    S100 calcium binding protein B
  • Antigen symbol:
    S100B
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    C-Terminal
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Antigen synonyms:
    S100beta|protein S100-B|S-100 calcium-binding protein|beta neural|NEF|S100-B|S100|S-100 protein subunit beta|S100 calcium-binding protein|beta chain
  • Amino acid number:
    64 to 78
  • UniProtKB:
    P04271
  • Sequence:
    DGDGECDFQEFMAFV
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    A synthetic peptide corresponding to a sequence at the C-terminus of human S100 beta (64-78aa DGDGECDFQEFMAFV), identical to the related rat and mouse sequences.
  • Purification:
    purified by immunogen affinity purification
  • Size:
    100 µg
  • Pk:
    0,1 mg

Specifications

About this item

Rabbit IgG polyclonal antibody for Protein S100-B(S100B) detection. Tested with WB in Human;Mouse;Rat.

S100 calcium binding protein B or S100B is a protein of the S-100 protein family. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21; however, this gene is located at 21q22.3. S100B is a glial-derived protein that is a well-established biomarker for severity of neurological injury and prognosis for recovery. S100 beta is a calcium-binding protein that is expressed at high levels in brain primarily by astrocytes. Addition of the disulfide-bonded dimeric form of S100 beta to primary neuronal and glial cultures and established cell lines induces axonal extension and alterations in astrocyte proliferation and phenotype, but evidence that S100 beta exerts the same effects in vivo has not been presented. Reeves et al.(1994) demonstrated that the same effects of the S100B protein are exerted in vivo. They found that both astrocytosis and neurite proliferation occurred in transgenic mice expressing elevated levels of S100b. They suggested that these transgenic mice represent a useful model for studies of the role of S100B in glial-neuronal interactions in normal development and function of the brain and for analyzing the significance of elevated levels of the protein in Down syndrome and Alzheimer disease.

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg Thimerosal, 0.05mg NaN3.

Tested Species: In-house tested species with positive results.
Predicted Species: Species predicted to be fit for the product based on sequence similarities.
Other applications have not been tested.
Optimal dilutions should be determined by end users.

Type: Primary
Antigen: S100 beta
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat