Specifications
- Antibody type:Primary
- Antigen name:Calbindin 2
- Antigen symbol:CALB2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:N-Terminal
- Cross adsorption:No
- Form:Liquid
- Antigen synonyms:29kD|CR|CAB29|calretinin|29 kDa Calbindin|Calbindin 2|Calbindin D29K|CAL2
- Amino acid number:4 to 18
- UniProtKB:P22676
- Sequence:PQQQPPYLHLAELTA
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the N-terminus of human Calretinin (4-18aa PQQQPPYLHLAELTA), identical to the related rat sequence.
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for Calretinin(CALB2) detection. Tested with WB, IHC-P in Human;Mouse;Rat.
Calbindin is a calcium-binding protein belonging to the troponin C superfamily. Calretinin is expressed in central and peripheral nervous system and in many normal and pathological tissues. The rat and human calretinin exhibit 98% sequence homology and 91% homology to many other species. Two calcium binding proteins, calbindin and calretinin, have been reported to be expressed in abundance in Purkinje cells and other cell types in the cerebellum.
Each vial contains 50% glycerol, 0.9mg NaCl, 0.2mg Na2HPO4.
Tested Species: In-house tested species with positive results.
Predicted Species: Species predicted to be fit for the product based on sequence similarities.
By Heat: Boiling the paraffin sections in 10mM citrate buffer, pH6.0, for 20mins is required for the staining of formalin/paraffin sections.
Other applications have not been tested.
Optimal dilutions should be determined by end users.
Type: Primary
Antigen: Calretinin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat