Anti-alpha-Actinin Mouse Monoclonal Antibody [clone: SA-20]
BSBTMA1104
- Antibody type:Primary
- Antigen name:alpha Actinin
- Antigen symbol:ACTN1
- Clonality:Monoclonal
- Clone:SA-20
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Rabbit,Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Ascites
- Form:Lyophilised
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:Rabbit skeletal alpha-actinin
- Size:100 µg
- Pk:0,1 mg
Mouse IgG monoclonal antibody for alpha-Actinin, actinin, alpha 2 (ACTN2) detection. Tested with WB, IHC-P, IHC-F in Human;mouse;rat. No cross reactivity with other proteins.
Reconstitute with 1,2% sodium acetate or neutral PBS.
If 1 ml of PBS is used, the antibody concentration will be 100 µg/ml.
Type: Primary
Antigen: alpha-Actinin
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Rabbit