Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-Vimentin Mouse Monoclonal Antibody [clone: VMT-24]
Anti-Vimentin Mouse Monoclonal Antibody [clone: VMT-24]
  BSBTMA1102
undefined
Anti-Vimentin Mouse Monoclonal Antibody [clone: VMT-24]
  BSBTMA1102

 

  • Antibody type:
    Primary
  • Antigen name:
    Vimentin
  • Antigen symbol:
    VIM
  • Clonality:
    Monoclonal
  • Clone:
    VMT-24
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Pig,
    Rabbit,
    Human,
    Rat
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Ascites
  • Form:
    Lyophilised
  • Antigen synonyms:
    CTRCT30|HEL113
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    Pig eye lens vimentin
  • Size:
    100 µg
  • Pk:
    0,1 mg

 

 

Mouse IgG monoclonal antibody for Vimentin, vimentin (VIM) detection. Tested with WB, IHC-P in Human;rat. No cross reactivity with other proteins.

Reconstitute with 1,2% sodium acetate or neutral PBS.

If 1 ml of PBS is used, the antibody concentration will be 100 µg/ml.

Type: Primary
Antigen: Vimentin
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rabbit, Pig, Rat