Anti-Tau Mouse Monoclonal Antibody [clone: TAU-93]
BSBTMA1093
- Antibody type:Primary
- Antigen name:Microtubule Associated Protein Tau
- Antigen symbol:MAPT
- Clonality:Monoclonal
- Clone:TAU-93
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human,Bovine
- Western blot:Yes
- Cross adsorption:No
- Format:Ascites
- Form:Lyophilised
- Antigen synonyms:PPP1R103|FTDP-17|TAU|PPND|MAPTL|MTBT1|MTBT2|MSTD|DDPAC
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:Bovine microtubule-associated proteins (MAPs)
- Size:100 µg
- Pk:0,1 mg
Mouse IgG monoclonal antibody for Tau, microtubule-associated protein tau (MAPT) detection. Tested with WB, IHC-P in Human;mouse;rat. No cross reactivity with other proteins.
Reconstitute with 1,2% sodium acetate or neutral PBS.
If 1 ml of PBS is used, the antibody concentration will be 100 µg/ml.
Type: Primary
Antigen: Tau
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Bovine