- Antibody type:Primary
- Antigen name:S100 calcium-binding protein B
- Antigen symbol:S100B
- Clonality:Monoclonal
- Clone:SA-12
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Rabbit,Human,Rat
- Western blot:Yes
- Cross adsorption:No
- Format:Ascites
- Form:Lyophilized
- Gene ID:Human: 6285, Mouse: 20203, Rat: 25742
- Antigen synonyms:Protein S100-B
- UniProtKB:Human: P04271, Mouse: P50114, Rat: P04631
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:Bovine brain S-100b.
- Size:100 µg
- Pk:0,1 mg
Mouse IgG monoclonal antibody for S-100 (beta-subunit), S100 calcium binding protein B (S100B) detection. Tested with IHC-P in Human;mouse;rat. No cross reactivity with other proteins.
S100 protein is a 21,000-Da component first isolated from brain by Moore(1965). Immunocytochemical studies demonstrated that S100 protein is produced by a wide variety of normal and neoplastic cells of mesodermal, neuroectodermal, and epithelial origin(Herrera et al., 1988). The S100 molecule is a dimer; thus there are 3 forms of S100: alpha-alpha, known as S-100a(0); alpha-beta, known as S-100a; and beta-beta, known as S-100b. S100 genes are clustered on human chromosome 1q21. S-100 protein expression by primary and metastatic adenocarcinomas.
1.2% sodium acetate or neutral PBS. If 1ml of PBS is used, the antibody concentration will be 100ug/ml.
Mouse ascites fluid, 1.2% sodium acetate, 2mg BSA, with 0.01mg NaN3 as preservative.
Other applications have not been tested.
Optimal dilutions should be determined by end users.
Type: Primary
Antigen: S-100 (beta-subunit)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rabbit, Rat