Anti-Myosin (Skeletal, Slow) Mouse Monoclonal Antibody [clone: IML-64]
Catalog # BSBTMA1064
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Myosin (Skeletal, Slow)
- Clonality:Monoclonal
- Clone:IML-64
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Rabbit,Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Ascites
- Form:Liquid
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:Human skeletal muscle myosin purified from myofibrils.
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: Myosin (Skeletal, Slow)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Rabbit