Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-MAP1 Mouse Monoclonal Antibody [clone: MP-1]
Anti-MAP1 Mouse Monoclonal Antibody [clone: MP-1]
Catalog # BSBTMA1056
Supplier:  Boster Bio
undefined
Anti-MAP1 Mouse Monoclonal Antibody [clone: MP-1]
Catalog # BSBTMA1056
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Modulator of Apoptosis 1
  • Antigen symbol:
    MAP1
  • Clonality:
    Monoclonal
  • Clone:
    MP-1
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Ascites
  • Form:
    Lyophilised
  • Antigen synonyms:
    FLJ77111|MAP1|MTAP1A|MAP1L|MAP1A
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    Rat brain microtubule-associated proteins (MAPs)
  • Size:
    100 µg
  • Pk:
    0,1 mg

Specifications

About this item

Mouse IgG monoclonal antibody for MAP1, microtubule-associated protein 1A (MAP1A) detection. Tested with WB, IHC-P, IHC-F in Rat. No cross reactivity with other proteins.

Reconstitute with 1,2% sodium acetate or neutral PBS.

If 1 ml of PBS is used, the antibody concentration will be 100 µg/ml.

Type: Primary
Antigen: MAP1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse, Rat