Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-GAP43 Mouse Monoclonal Antibody [clone: GAP-8A12]
Anti-GAP43 Mouse Monoclonal Antibody [clone: GAP-8A12]
Catalog # BSBTMA1042
Supplier:  Boster Bio
undefined
Anti-GAP43 Mouse Monoclonal Antibody [clone: GAP-8A12]
Catalog # BSBTMA1042
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Growth associated protein 43
  • Antigen symbol:
    GAP43
  • Clonality:
    Monoclonal
  • Clone:
    GAP-8A12
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG2a
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Ascites
  • Form:
    Lyophilised
  • Antigen synonyms:
    axonal membrane protein GAP-43|growth-associated protein 43|neuron growth-associated protein 43 protein F1|neuromodulin|nerve growth-related peptide GAP43|calmodulin-binding protein P-57|neural phosphoprotein B-50|PP46|B-50
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    GAP-43 from neonatal rat forebrain membranes
  • Size:
    100 µg
  • Pk:
    0,1 mg

Specifications

About this item

Mouse IgG monoclonal antibody for GAP43, growth associated protein 43 (GAP43) detection. Tested with WB, IHC-P, IHC-F in Human; mouse; rat; chicken; snake. No cross reactivity with otherproteins.

Reconstitute with 1,2% sodium acetate or neutral PBS.

If 1 ml of PBS is used, the antibody concentration will be 100 µg/ml.

Type: Primary
Antigen: GAP43
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat