Anti-Cyclin A Mouse Monoclonal Antibody [clone: CY-28]
BSBTMA1032
- Antibody type:Primary
- Antigen name:Cyclin A
- Antigen symbol:CCNA1
- Clonality:Monoclonal
- Clone:CY-28
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a
- Reactivity:Human,Bovine,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Ascites
- Form:Lyophilised
- Antigen synonyms:CCN1|CCNA
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:Recombinant bovine cyclin A
- Size:100 µg
- Pk:0,1 mg
Mouse IgG monoclonal antibody for Cyclin A, cyclin A2 (CCNA2) detection. Tested with WB, ICC in Human;mouse. No cross reactivity with other proteins.
Reconstitute with 1,2% sodium acetate or neutral PBS.
If 1 ml of PBS is used, the antibody concentration will be 100 µg/ml.
Type: Primary
Antigen: Cyclin A
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Bovine