Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-Cathepsin D Mouse Monoclonal Antibody [clone: CP-D13]
Anti-Cathepsin D Mouse Monoclonal Antibody [clone: CP-D13]
Catalog # BSBTMA1013
Supplier:  Boster Bio
undefined
Anti-Cathepsin D Mouse Monoclonal Antibody [clone: CP-D13]
Catalog # BSBTMA1013
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Cathepsin D
  • Antigen symbol:
    CTSD
  • Clonality:
    Monoclonal
  • Clone:
    CP-D13
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG2a
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Ascites
  • Form:
    Lyophilised
  • Antigen synonyms:
    lysosomal aspartyl protease|HEL-S-130P|ceroid-lipofuscinosis|lysosomal aspartyl peptidase|CPSD|CLN10|neuronal 10
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    Human liver cathepsin D
  • Size:
    100 µg
  • Pk:
    0,1 mg

Specifications

About this item

Mouse IgG monoclonal antibody for Cathepsin D, cathepsin D (CTSD) detection. Tested with WB, IHC-P in Human. No cross reactivity with other proteins.

Reconstitute with 1,2% sodium acetate or neutral PBS.

If 1 ml of PBS is used, the antibody concentration will be 100 µg/ml.

Type: Primary
Antigen: Cathepsin D
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human