Anti-Calmodulin Mouse Monoclonal Antibody [clone: CM-16]
Catalog # BSBTMA1010
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Calmodulin
- Antigen symbol:CAM
- Clonality:Monoclonal
- Clone:CM-16
- Conjugation:KLH (Keyhole Limpet Hemocyanin)
- Host:Mouse
- Isotype:IgG1
- Reactivity:Chicken,Rat
- Western blot:Yes
- Cross adsorption:No
- Format:Ascites
- Form:Lyophilized
- Antigen synonyms:CALM3|CAM2|CAMB|CALML2|CALM2|CAMIII|DD132|CAM3|CAM|CAMC|CaM|CAMI|CAM1|PHKD|CALM1|CALM
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:Preparation of purified calmodulin from Dictyostelium discoideum conjugated to KLH.
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: Calmodulin
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Chicken, Rat