Anti-Calcineurin alpha Mouse Monoclonal Antibody [clone: CC-6]
Catalog # BSBTMA1008
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Calcineurin A alpha
- Antigen symbol:CALNA
- Clonality:Monoclonal
- Clone:CC-6
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human,Bovine,Rat
- Western blot:Yes
- Cross adsorption:No
- Format:Ascites
- Form:Liquid
- Gene ID:Human: 5534, Mouse: 19058, Rat: 29748
- Antigen synonyms:CALN|PPP2B|CNA|CNA1|CALNA1|CCN1 |EC 3.1.3.16|CALNA
- UniProtKB:Human: P63098, Mouse: Q63810, Rat: P63100
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:Bovine brain calcineurin.
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Mouse IgG monoclonal antibody for Calcineurin alpha, protein phosphatase 3, regulatory subunit B, alpha (PPP3R1) detection. Tested with WB, IHC-P in Human;rat. No cross reactivity with other proteins.
- Calcineurin, the Ca(2+)/calmodulin-regulated protein phosphatase, first detected in skeletal muscle and brain, has been found in all cells from yeast to mammals. Calcineurin A alpha(PPP3CA),is located on human chromosomes 4, Chromosomal mapping of the human genes for the calmodulin-dependent protein phosphatase(calcineurin) catalytic subunit. Calcineurin regulates bone formation by the osteoblast.
Mouse ascites fluid, 1.2% sodium acetate, 2mg BSA, with 0.01mg NaN3 as preservative.
Other applications have not been tested.
Optimal dilutions should be determined by end users.
Type: Primary
Antigen: Calcineurin alpha
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat, Bovine