Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-Calbindin-D Mouse Monoclonal Antibody [clone: CB-D7]
Anti-Calbindin-D Mouse Monoclonal Antibody [clone: CB-D7]
  BSBTMA1007
undefined
Anti-Calbindin-D Mouse Monoclonal Antibody [clone: CB-D7]
  BSBTMA1007

 

  • Antibody type:
    Primary
  • Antigen name:
    Calbindin 1 28kDa
  • Antigen symbol:
    CALB1
  • Clonality:
    Monoclonal
  • Clone:
    CB-D7
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human,
    Bovine,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Ascites
  • Form:
    Lyophilised
  • Antigen synonyms:
    CALB|Calb|Calb-1|CB|Calbindin 1 28kDa|Brain-2
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    Bovine kidney calbindin-D
  • Size:
    100 µg
  • Pk:
    0,1 mg

 

 

Mouse IgG monoclonal antibody for Calbindin-D, calbindin 1, 28 kDa (CALB1) detection. Tested with WB, IHC-P in Human;mouse;rat;bovine. No cross reactivity with other proteins.

Reconstitute with 1,2% sodium acetate or neutral PBS.

If 1 ml of PBS is used, the antibody concentration will be 100 µg/ml.

Type: Primary
Antigen: Calbindin-D
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Bovine