Anti-AFP Mouse Monoclonal Antibody [clone: AP-1]
BSBTMA1001
- Antibody type:Primary
- Antigen name:Alpha 1 Fetoprotein Receptor
- Antigen symbol:AFP
- Clonality:Monoclonal
- Clone:AP-1
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a
- Reactivity:Human
- Cross adsorption:No
- Format:Ascites
- Form:Lyophilised
- Antigen synonyms:FETA|Alpha-1-Fetoprotein|HPAFP
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:Human alpha-fetoprotein
- Size:100 µg
- Pk:0,1 mg
Mouse IgG monoclonal antibody for AFP, alpha-fetoprotein (AFP) detection. Tested with IHC-P in Human. No cross reactivity with other proteins.
Reconstitute with 1,2% sodium acetate or neutral PBS.
If 1 ml of PBS is used, the antibody concentration will be 100 µg/ml.
Type: Primary
Antigen: AFP
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human