Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-C20orf26 Rabbit Polyclonal Antibody
Anti-C20orf26 Rabbit Polyclonal Antibody
Catalog # ABCAAB121300-100
Supplier:  Abcam
Anti-C20orf26 Rabbit Polyclonal Antibody
Catalog # ABCAAB121300-100
Supplier:  Abcam

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Chromosome 20 open reading frame 26
  • Antigen symbol:
    C20orf26
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

Caution:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: C20orf26 (chromosome 20 open reading frame 26)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human