Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-Complex I subunit 8 kDa Mouse Monoclonal Antibody [clone: 17C8E4E11]
Anti-Complex I subunit 8 kDa Mouse Monoclonal Antibody [clone: 17C8E4E11]
Catalog # ABCAAB110245-100
Supplier:  Abcam
Anti-Complex I subunit 8 kDa Mouse Monoclonal Antibody [clone: 17C8E4E11]
Catalog # ABCAAB110245-100
Supplier:  Abcam

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Complex I subunit 8 kDa
  • Clonality:
    Monoclonal
  • Clone:
    17C8E4E11
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: Complex I subunit 8 kDa
Clonality: monoclonal
Clone: 17C8E4E11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: