Order Entry
Northern Ireland
ContactUsLinkComponent
 
Anti-Myosin Light Chain 2 Rabbit Polyclonal Antibody
Catalog # ABCAAB79935-100
Supplier:  Abcam
undefined
Anti-Myosin Light Chain 2 Rabbit Polyclonal Antibody
Catalog # ABCAAB79935-100
Supplier:  Abcam

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    MLC2
  • Antigen symbol:
    Myosin Light Chain 2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Seq:
    1286
  • Form:
    Liquid
  • Antigen synonyms:
    MYL2|MLC-2|MLC-2v|Cardiac myosin light chain 2|Ventricular myosin light chain 2|MLC-2s/v
  • Molecular weight:
    18789 Da
  • Storage temperature:
    Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C or –80 °C. Avoid freeze / thaw cycle.
  • Concentration:
    0,7 mg/ml
  • Shipping temperature:
    4 °C
  • Immunogen:
    The exact immunogen used to generate this antibody is proprietary information.
  • Tested applications:
    IHC-P
  • Purification:
    Immunogen Affinity purification
  • Size:
    100 µg
  • Pack type:
    Vial
  • Pk:
    100 µG

Specifications

About this item

Rabbit polyclonal to Myosin Light Chain 2.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Myosin Light Chain 2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat