Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-E2F2 Mouse Monoclonal Antibody [clone: 2302C4a]
Anti-E2F2 Mouse Monoclonal Antibody [clone: 2302C4a]
  ABCAAB70731-100
 :  Abcam
undefined
Anti-E2F2 Mouse Monoclonal Antibody [clone: 2302C4a]
  ABCAAB70731-100
 :  Abcam

 

  • Antibody type:
    Primary
  • Antigen name:
    E2F transcription factor 2
  • Antigen symbol:
    E2F2
  • Clonality:
    Monoclonal
  • Clone:
    2302C4a
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG2a
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pack type:
    Vial
  • Pk:
    100 µG

 

 

Mouse monoclonal [2302C4a] to E2F2.

 : All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: E2F2
Clonality: Monoclonal
Clone: 2302C4a
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: