Anti-Tissue Plasminogen Activator Rabbit Polyclonal Antibody
Catalog # ABCAAB62763-200
Supplier: Abcam
Specifications
- Antibody type:Primary
- Antigen name:Plasminogen Activator, Tissue
- Antigen symbol:PLAT
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western blot:Yes
- Seq:1358
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 µg
- Pack type:Vial
- Pk:200 µG
Specifications
About this item
Rabbit polyclonal to Tissue Plasminogen Activator.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: Tissue Plasminogen Activator
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Rat