Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-Properdin/PFC Mouse Monoclonal Antibody [clone: 10-18]
Anti-Properdin/PFC Mouse Monoclonal Antibody [clone: 10-18]
Catalog # ABCAAB58984-50
Supplier:  Abcam
undefined
Anti-Properdin/PFC Mouse Monoclonal Antibody [clone: 10-18]
Catalog # ABCAAB58984-50
Supplier:  Abcam

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    Properdin
  • Clonality:
    Monoclonal
  • Clone:
    10-18
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Seq:
    1202
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 µg
  • Pack type:
    Vial
  • Pk:
    50 µG

Specifications

About this item

Caution:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Properdin
Clonality: monoclonal
Clone: 42295
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human