Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-CAD Rabbit Monoclonal Antibody [clone: EP710Y]
Anti-CAD Rabbit Monoclonal Antibody [clone: EP710Y]
Supplier:  Abcam
New Product
Certificates
undefined
Anti-CAD Rabbit Monoclonal Antibody [clone: EP710Y]
Supplier:  Abcam
New Product

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Catalog No:
  • ABCAAB40800-100
  • ABCAAB40800-1ML
  • Antigen symbol:
  • CAD
  • Antigen name:
  • carbamoyl-phosphate synthetase 2, aspartate transcarbamylase, and dihydroorotase
  • Antibody type:
  • Primary
  • Clonality:
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Clone:
  • EP710Y
  • Host:
  • Rabbit
  • Isotype:
  • IgG
  • Western blot:
  • Yes
  • ImmunoChemistry:
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Flow cytometry:
  • Yes
  • Storage temperature:
  • Cool
  • Shipping temperature:
  • Ice

Specifications

Frequently Bought Together