Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-CAD Rabbit Monoclonal Antibody [clone: EP711Y]
Anti-CAD Rabbit Monoclonal Antibody [clone: EP711Y]
  ABCAAB40765-100
 :  Abcam
undefined
Anti-CAD Rabbit Monoclonal Antibody [clone: EP711Y]
  ABCAAB40765-100
 :  Abcam

 

  • Antibody type:
    Primary
  • Antigen name:
    carbamoyl-phosphate synthetase 2, aspartate transcarbamylase, and dihydroorotase
  • Antigen symbol:
    CAD
  • Clonality:
    Monoclonal
  • Clone:
    EP711Y
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µl
  • Pack type:
    Vial
  • Pk:
    100 µl

 

 

Rabbit monoclonal [EP711Y] to CAD.

 :

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: CAD (carbamoyl-phosphate synthetase 2, aspartate transcarbamylase, and dihydroorotase)
Clonality: monoclonal
Clone: EP711Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: