Anti-C3a/C3a des Arg Mouse Monoclonal Antibody [clone: K13/16]
Catalog # ABCAAB36385-100
Supplier: Abcam
Specifications
- Antibody type:Primary
- Antigen name:complement component 3
- Antigen symbol:C3
- Clonality:Monoclonal
- Clone:013-16
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pack type:Vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [K13/16] to C3a / C3a des Arg.
Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: C3a / C3a des Arg
Clonality: Monoclonal
Clone: K13/16
Conjugation:
Epitope: Recognizes an epitope present on human C3, C3a and C3a(desArg).
Host: Mouse
Isotype: IgG1
Reactivity: Human