- Antibody type:Primary
- Antigen name:tumor necrosis factor receptor superfamily, member 4
- Antigen symbol:TNFRSF4
- Clonality:Monoclonal
- Clone:MRC OX-40
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Rat
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pack type:Vial
- Pk:50 µG
FITC Mouse monoclonal [MRC OX-40] to CD134 / OX40L receptor.
All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: TNFRSF4 (tumor necrosis factor receptor superfamily, member 4)
Clonality: monoclonal
Clone: MRC OX-40
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Rat