Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-Proteasome 20S alpha 1+2+3+5+6+7 Mouse Monoclonal Antibody [clone: MCP231]
Anti-Proteasome 20S alpha 1+2+3+5+6+7 Mouse Monoclonal Antibody [clone: MCP231]
Catalog # ABCAAB22674-50
Supplier:  Abcam
undefined
Anti-Proteasome 20S alpha 1+2+3+5+6+7 Mouse Monoclonal Antibody [clone: MCP231]
Catalog # ABCAAB22674-50
Supplier:  Abcam

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    Proteasome 20S alpha 1+2+3+5+6+7
  • Clonality:
    Monoclonal
  • Clone:
    MCP231
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 µl
  • Pack type:
    Vial
  • Pk:
    50 µl

Specifications

About this item

Mouse monoclonal [MCP231] to Proteasome 20S alpha 1+2+3+5+6+7.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Proteasome 20S alpha 1+2+3+5+6+7
Clonality: Monoclonal
Clone: MCP231
Conjugation:
Epitope: Reacts with the peptide sequence TVWSPQGRLHQVEYAMEA encompassing the prosbox I motif common to alpha type subunits, although not necessarily identical in all. In general this motif is phylogenetically preserved.
Host: Mouse
Isotype: IgG1
Reactivity: Human