Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-PRKD1 Rabbit Polyclonal Antibody
Anti-PRKD1 Rabbit Polyclonal Antibody
Supplier:  Biorbyt
Certificates
undefined
Anti-PRKD1 Rabbit Polyclonal Antibody
Supplier:  Biorbyt

Specifications

  • Catalog No:
  • BIRBORB99165-20
  • BIRBORB99165-100
  • Antigen symbol:
  • PRKD1
  • PRKD1
  • Antigen name:
  • Protein kinase D1
  • Protein kinase D1
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human, Rat, Mouse
  • Human, Rat, Mouse
  • Host:
  • Rabbit
  • Rabbit
  • Isotype:
  • IgG
  • IgG
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Modification:
  • phospho-Ser738, phospho-Ser742
  • phospho-Ser738, phospho-Ser742
  • Immunogen:
  • A synthesized peptide derived from human PKD1/2/3/PKC around the phosphorylation site of Serine 738+Serine 742 .
  • A synthesized peptide derived from human PKD1/2/3/PKC around the phosphorylation site of Serine 738+Serine 742 .
  • Molecular weight:
  • 101 kDa
  • 101 kDa
  • Purification:
  • purified by immunoaffinity chromatography
  • purified by immunoaffinity chromatography
  • Concentration:
  • 20 ug: 0,2 mg/ml; 50 ug: 0,5 mg/ml; 100 ug: 1 ug/ul; 200 ug: 1 ug/ul
  • 20 ug: 0,2 mg/ml; 50 ug: 0,5 mg/ml; 100 ug: 1 ug/ul; 200 ug: 1 ug/ul
  • Storage buffer:
  • Phosphate buffer saline, pH. 7.4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
  • Phosphate buffer saline, pH. 7.4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information

Frequently Bought Together