Anti-C-Peptide Rabbit Polyclonal Antibody
Catalog # BIRBORB98717-1
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:C-peptide
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Type: Primary
Antigen: C-Peptide
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: