Anti-alphaT-Catenin Mouse Monoclonal Antibody
Catalog # BIRBORB98231-0.1
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:alpha T-Catenin
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Reactivity:Human
- Western blot:Yes
- Storage buffer:PBS containing 0,09% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: alphaT-Catenin
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human