Anti-lambda light chainlambda light chain Sheep Polyclonal Antibody
Catalog # BIRBORB98175-10
Supplier: Biorbyt
Specifications
- Antibody type:Secondary
- Antigen name:lambda light chain
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Sheep
- Isotype:IgG
- Reactivity:Mouse
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:10 mg
- Pk:10 mg
Specifications
About this item
Type: Secondary
Antigen: Mouse Ig lambda
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Mouse