Anti-lambda light chainlambda light chain Rabbit Polyclonal Antibody
Catalog # BIRBORB98169-1
Supplier: Biorbyt
Specifications
- Antibody type:Secondary
- Antigen name:lambda light chain
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:1 ml
- Pk:1 mL
Specifications
About this item
Type: Secondary
Antigen: human Ig lambda
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human