Anti-Malic dehydrogenase Rabbit Polyclonal Antibody
BIRBORB98143-1
: Biorbyt
- Antibody type:Primary
- Antigen name:Malate Dehydrogenase
- Antigen symbol:MDH
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Pig
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:1 mg
- Pk:1 mg
Type: Primary
Antigen: Malic dehydrogenase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Pig