Anti-GPR151 Rabbit Polyclonal Antibody
Catalog # BIRBORB95457-50
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:G Protein-Coupled Receptor 151
- Antigen symbol:GPR151
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Storage buffer:PBS, pH 7,4, 150 mM sodium chloride, 0,02% sodium azide, 50% glycerol
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
ELISA: 1:1000; Immunofluorescence: 1:100-1:500; Immunochemistry: 5 ?g/ml; Western Blot: 1:500-1:1000
Type: Primary
Antigen: GPR151 (G protein-coupled receptor 151)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat