Anti-C Kit Mouse Monoclonal Antibody
Catalog # BIRBORB95056-50
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Proto-oncogene Receptor Tyrosine Kinase
- Antigen symbol:KIT
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b kappa
- Reactivity:Human
- Western blot:Yes
- Storage buffer:PBS, pH 7,2
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Immunochemistry: 2.5 ?g/ml; Western Blot: 1:500-1:1000
Type: Primary
Antigen: C Kit
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human