Anti-SP140 Mouse Monoclonal Antibody
Catalog # BIRBORB95035-50
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:SP140 nuclear body Protein
- Antigen symbol:SP140
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Storage buffer:PBS, pH 7,2
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Immunochemistry: 5 ?g/ml
Type: Primary
Antigen: SP140 (SP140 nuclear body protein)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human