Anti-SLC40A1 Rabbit Polyclonal Antibody
BIRBORB87684-50
: Biorbyt
- Antibody type:Primary
- Antigen name:solute carrier family 40 (iron-regulated transporter), member 1
- Antigen symbol:Solute carrier family 40, member 1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Western blot:Yes
- Storage buffer:PBS, 0,01% sodium azide, 50% glycerol.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
ELISA: 1 ?g/ml; Immunochemistry: 20 ?g/ml; Western Blot: 1 ?g/ml
Type: Primary
Antigen: SLC40A1 (solute carrier family 40 (iron-regulated transporter), member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: