Anti-HTR2B Rabbit Polyclonal Antibody
Catalog # BIRBORB85370-50
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:5-hydroxytryptamine (serotonin) receptor 2B
- Antigen symbol:HTR2B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Storage buffer:PBS, 0,1% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Synthetic 17 amino acid peptide from 3rd cytoplasmic domain of human 5HT2B Receptor
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Immunochemistry: 2 ?g/ml
Type: Primary
Antigen: HTR2B (5-hydroxytryptamine (serotonin) receptor 2B, G protein-coupled)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human