Anti-coiled coil domain containing 102B Rabbit Polyclonal Antibody
BIRBORB74271-100
: Biorbyt
- Antibody type:Primary
- Antigen name:coiled coil domain containing 102B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Storage temperature:Cool
- Concentration:0,97 mg/ml
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: coiled coil domain containing 102B
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human