Anti-MPP6 Rabbit Polyclonal Antibody
Catalog # BIRBORB73598-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Membrane Protein, Palmitoylated 6 (MAGUK p55 subfamily member 6)
- Antigen symbol:MPP6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Storage temperature:Cool
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: MPP6 (membrane protein, palmitoylated 6 (MAGUK p55 subfamily member 6))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human