Anti-RXFP2 Rabbit Polyclonal Antibody
BIRBORB137404-50
: Biorbyt
- Antibody type:Primary
- Antigen name:Relaxin/insulin-like family peptide receptor 2
- Antigen symbol:RXFP2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Storage buffer:0,1 M Tris-glycine, pH 7, 10% Glycerol, 0,01% Thimerosal
- Storage temperature:Cool
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Immunogen:RXFP2/LGR8 antibody was raised against synthetic peptide corresponding to a region within amino acids 690 and 754 of Relaxin Receptor 2 (SwissProt Q8WXD0). Percent identity by BLAST analysis: Human, Monkey (100%).
- Purification:purified by immunoaffinity chromatography
- Size:50 µl
- Pk:50 µl
Immunochemistry: 10 ?g/ml
Type: Primary
Antigen: RXFP2 (relaxin/insulin-like family peptide receptor 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: