Anti-RHOU Rabbit Polyclonal Antibody
Catalog # BIRBORB137392-50
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:ras homolog family member U
- Antigen symbol:RHOU
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Amino acid number:1 to 11
- Storage buffer:PBS, pH 7, 10% Glycerol, 0,01% Thimerosal
- Storage temperature:Cool
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Immunogen:ARHU/RHOU antibody was raised against synthetic peptide contain a sequence corresponding to a region within amino acids 1 and 11 (Q7L0Q8) of WRCH1. Percent identity by BLAST analysis: Human (100%).
- Purification:purified by immunoaffinity chromatography
- Size:50 µl
- Pk:50 µl
Specifications
About this item
Immunochemistry: 10 ?g/ml
Type: Primary
Antigen: RHOU (ras homolog family member U)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human