Anti-NOX4 Rabbit Polyclonal Antibody (Cy7®)
Catalog # BIRBORB123803-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:NADPH oxidase 4
- Antigen symbol:NOX4
- Clonality:Polyclonal
- Conjugation:Cy7®
- Host:Rabbit
- Isotype:IgG
- Form:liquid
- Storage buffer:10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- Molecular weight:64 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Synthetic peptide
- Purification:purified by affinity chromatography
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: NOX4 (NADPH oxidase 4)
Clonality: polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: