Anti-MT-ND6 Rabbit Polyclonal Antibody (Cy7®)
Catalog # BIRBORB123784-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:mitochondrially encoded NADH dehydrogenase 6
- Antigen symbol:MT-ND6
- Clonality:Polyclonal
- Conjugation:Cy7®
- Host:Rabbit
- Isotype:IgG
- Form:liquid
- Storage buffer:10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- Molecular weight:19 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:KLH conjugated synthetic peptide derived from human MTND6.
- Purification:purified by affinity chromatography
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: MT-ND6 (mitochondrially encoded NADH dehydrogenase 6)
Clonality: polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: