Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-PH Domain-containing family E member 1 Rabbit Polyclonal Antibody
Anti-PH Domain-containing family E member 1 Rabbit Polyclonal Antibody
Catalog # BIRBORB109563-100
Supplier:  Biorbyt
Anti-PH Domain-containing family E member 1 Rabbit Polyclonal Antibody
Catalog # BIRBORB109563-100
Supplier:  Biorbyt

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    PH Domain-containing family E member 1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Immunogen:
    Synthetic peptide derived from the human PHLPP1 protein
  • Purification:
    purified by antigen immunoaffinity chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: PH Domain-containing family E member 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human