Anti-Influenza A Neuraminidase N1 Rabbit Polyclonal Antibody
Catalog # BIRBORB109500-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Influenza A Neuraminidase N1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Synthetic peptide corresponding to the neuraminidase protein from seasonal Influenza A/Georgia/20/2006 (H1N1) (Genbank accession no. ACA33620)
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: Influenza A Neuraminidase N1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human