Anti-Influenza A Hemagglutinin H5 Mouse Monoclonal Antibody
Catalog # BIRBORB109481-500
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Influenza A Hemagglutinin H5
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Purified avian influenza virus type A (H5N1)
- Purification:purified by Protein G chromatography
- Size:500 µg
- Pk:500 µG
Specifications
About this item
Type: Primary
Antigen: Influenza A Hemagglutinin H5
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: