Anti-SUMO1 Rabbit Polyclonal Antibody
Catalog # BIRBORB109440-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:small ubiquitin-like modifier 1
- Antigen symbol:Small ubiquitin-related modifier 1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Synthetic peptide derived from the human SUMO-1 protein.
- Purification:purified by ammonium sulfate precipitation
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: SUMO1 (small ubiquitin-like modifier 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human